Learn More
GSC Go Science Crazy Voltaic Cell with 8 Electrodes and Porous Cup
Helps to visually demonstrate electro-chemical changes taking place. The GSC Voltaic Cell with 8 Electrodes and Porous Cup is supplied with electrodes that includes; copper, aluminum, zinc, tin, brass, iron and nickel, round carbon rod and a porous cup. The Voltaic Cell is ideal for demonstrating the characteristics of a primary cell.
Manufacturer:  GSC Go Science Crazy 41826
Catalog No. S13611
Add to cart
Alert:
Not a toy. Product intended for ages 13 and up.
Includes: 7 flat 18 x 100mm electrodes (copper, aluminum, zinc, tin, brass, iron, nickel), 1 round carbon electrode, porous cup and manual
Description
Description
  • Supplied with 7 flat electrodes,
  • Transparent cup for easy viewing
  • Holds both round and flat electrodes
Specifications
Specifications
Demonstrating the characteristics of a primary cell
Voltaic cell
7 flat 18 x 100mm electrodes (copper, aluminum, zinc, tin, brass, iron, nickel), 1 round carbon electrode, porous cup and manual
Transparent