Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GSDMB Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554929
Description
Immunogen sequence: AGGDMIAVRS LVDADRFRCF HLVGEKRTFF GCRHYTTGLT LMDILDTDGD KWLDELDSGL QGQKAEFQIL DNVDSTGELI VRLPKEIT Highest antigen sequence identity to the following orthologs: Mouse - 34%, Rat - 33%.
This gene encodes a member of the gasdermin-domain containing protein family. Other gasdermin-family genes are implicated in the regulation of apoptosis in epithelial cells, and are linked to cancer. Multiple transcript variants encoding different isoforms have been found for this gene. Additional variants have been described, but they are candidates for nonsense-mediated mRNA decay (NMD) and are unlikely to be protein-coding.
Specifications
GSDMB | |
Polyclonal | |
Unconjugated | |
GSDMB | |
gasdermin B; gasdermin-B; Gasdermin-like protein; GSDMB; GSDML; PP4052; PRO2521 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55876 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8TAX9 | |
GSDMB | |
Recombinant protein corresponding to Human GSDMB. Recombinant protein control fragment (Product #RP-93176). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction