Learn More
Aviva Systems Biology Corp GSG1 Antibody - N-terminal region (ARP53801_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP53801_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GSG1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GSG1. Alternate names: MGC111023, MGC3146ApplicationWB. Species reactivity: Human, Mouse
Catalog No. 50-014-61132