Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSG2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$482.50

Specifications

Antigen GSG2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP179836
SDP
View Documents
Novus Biologicals
NBP179836
100 μL
Each of 1 for $482.50
Add to Cart
 
Description

Description

GSG2 Polyclonal specifically detects GSG2 in Human samples. It is validated for Western Blot.
Specifications

Specifications

GSG2
Polyclonal
Rabbit
Cell Cycle and Replication
NP_114171
83903
Synthetic peptide directed towards the middle region of human GSG2The immunogen for this antibody is GSG2. Peptide sequence QEAVRREHQEASVPKGRIVPRGIDRLERTRSSRKSKHQEATETSLLHSHR.
Primary
Western Blot
Unconjugated
RUO
Human
EC 2.7.11.1, germ cell associated 2 (haspin), Germ cell-specific gene 2 protein, Haploid germ cell-specific nuclear protein kinase, HASPIN, H-haspin, serine/threonine-protein kinase haspin
GSG2
IgG
88 kDa
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit