Tocris™ GSK 1562590 hydrochloride

High affinity, selective urotensin II (UT) receptor antagonist

Manufacturer: Tocris™ 5110/10

 View more versions of this product

Catalog No. 51-101-0

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors GSK 1562590 hydrochloride
    Purity >98
    Molecular Weight 617.95
    Quantity 10mg
    Product Type GSK 1562590 hydrochloride

    High affinity and selective urotensin II (UT) receptor antagonist (pKi values are 9.14, 9.28, 9.34, 9.64 and 9.66 at monkey, human, mouse, cat and rat recombinant receptors respectively). Exhibits selectivity for UT receptors over a range of GPCRs, ion channels, enzymes and neurotransmitter transporters. Supresses human urotensin-II (hU-II)-induced contraction of isolated rat aorta in vitro and ex vivo. Inhibits the hU-II-induced increase in mean blood pressure in vivo. Orally active.

    • Chemical Name: N-[(1R)-1-[3'-(Aminocarbonyl)[1,1'-biphenyl]-4-yl]-2-(1-pyrrolidinyl)ethyl]-6,7-dichloro-2,3-dihydro-N-methyl-3-oxo-4H-1,4-benzoxazine-4-acetamide hydrochloride
    • Formula: C30H30Cl2N4O4.HCl
    • CAS Number: 1003878-07-6
    • Solubility: Soluble to 100mM in DMSO
    • Purity: >98

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit