Tocris™ GSK 2193874

Potent and selective TRPV4 inhibitor; orally active

Manufacturer: Tocris™ 5106/10

 View more versions of this product

Catalog No. 51-061-0

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors GSK 2193874
    Molecular Weight 691.62
    Quantity 10mg
    Product Type GSK 2193874

    Potent and selectiveTRPV4 inhibitor (IC50 values are 2 and 40nM for rat and human receptors, respectively); inhibits Ca2+ influx through TRPV4 channels. Prevents and reverses pulmonary edema after myocardial infarction in vivo models. Selective over a panel of ~200 human receptors, channels and enzymes. Orally active.

    • Chemical Name: 3-([1,4'-Bipiperidin]-1'-ylmethyl)-7-bromo-N-(1-phenylcyclopropyl)-2-[3-(trifluoromethyl)phenyl]-4-quinolinecarboxamide
    • Formula: C37H38BrF3N4O
    • CAS Number: 1336960-13-4
    • Solubility: Soluble to 20mM in 1eq. HCl and to 50mM in DMSO

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit