Tocris™ GSK 2194069

Potent human fatty acid synthase (hFASN) inhibitor

Manufacturer: Tocris™ 5303/10

 View more versions of this product

Catalog No. 53-031-0

  N/A
Add to Cart

Disclaimers:

For Research Use Only

  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Description GSK 2194069
    Quantity 10mg
    Product Type GSK 2194069
    Molecular Weight 428.48
    Purity >99

    Potent human fatty acid synthase (hFASN) inhibitor (IC50 = 7.7nM); inhibits hFAS β-ketoacyl reductase activity. Inhibits lipid synthesis and attenuates proliferation of A549 non-small-cell lung cancer cells (EC50 = 15nM) in vitro.

    • Chemical Name: 4-[4-(5-Benzofuranyl)phenyl]-5-[[(3S)-1-(cyclopropylcarbonyl)-3-pyrrolidinyl]methyl]-2,4-dihydro-3H-1,2,4-triazol-3-one
    • Formula: C25H24N4O3
    • CAS Number: 1332331-08-4
    • Solubility: Soluble to 100mM in DMSO and to 20mM in ethanol
    • Purity: >99

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit