Tocris™ GSK 2194069

Potent human fatty acid synthase (hFASN) inhibitor

Manufacturer: Tocris™ 5303/50

 View all options for this product

Catalog No. 53-035-0

  Check Availability
Add to Cart


For Research Use Only


    Description GSK 2194069
    Molecular Weight 428.48
    Purity >99
    Quantity 50mg
    Product Type GSK 2194069

    Potent human fatty acid synthase (hFASN) inhibitor (IC50 = 7.7nM); inhibits hFAS β-ketoacyl reductase activity. Inhibits lipid synthesis and attenuates proliferation of A549 non-small-cell lung cancer cells (EC50 = 15nM) in vitro.

    • Chemical Name: 4-[4-(5-Benzofuranyl)phenyl]-5-[[(3S)-1-(cyclopropylcarbonyl)-3-pyrrolidinyl]methyl]-2,4-dihydro-3H-1,2,4-triazol-3-one
    • Formula: C25H24N4O3
    • CAS Number: 1332331-08-4
    • Solubility: Soluble to 100mM in DMSO and to 20mM in ethanol
    • Purity: >99

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit