Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GSK 2239633A

Potent CCR4 antagonist

$240.50 - $1014.00

Specifications

CAS 1240516-71-5
Chemical Name or Material N-[[3-[[3-[[(5-Chloro-2-thienyl)sulfonyl]amino]-4-methoxy-1H-indazol-1-yl]methyl]phenyl]methyl]-2-hydroxy-2-methylpropanamide
Molecular Formula C24H25ClN4O5S2
Purity 98%
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
70-711-0
View Documents
Tocris Bioscience™
7071/10
10 mg
Each for $240.50
Add to cart
 
70-715-0
View Documents
Tocris Bioscience™
7071/50
50 mg
Each for $1,014.00
Add to cart
 
Description

Description

  • Potent CCR4 antagonist (pIC50 = 7.83 in a 35S-GTPγS competition assay).
  • Inhibits thymus activation-regulated chemokine (TARC) -induced F-actin polymerization in human whole blood.
Specifications

Specifications

1240516-71-5
N-[[3-[[3-[[(5-Chloro-2-thienyl)sulfonyl]amino]-4-methoxy-1H-indazol-1-yl]methyl]phenyl]methyl]-2-hydroxy-2-methylpropanamide
C24H25ClN4O5S2
98%
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit