Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris™ GSK 2830371

Potent and selective allosteric inhibitor of Wip1 phosphatase

Manufacturer:  Tocris™ 5140/50

 View more versions of this product

Catalog No. 51-405-0


  Check Availability
Add to cart

Disclaimers:

For Research Use Only


Specifications

Description

Potent and selective allosteric inhibitor of Wip1 phosphatase (IC50 = 6nM). Exhibits selectivity for Wip1 over 21 other phosphatases. Increases phosphorylation of Wip1 substrates, including p53, Chk2, H2AX and ATM. Attenuates tumor cell growth in a variety of lymphoid cell lines. Also inhibits lymphoma xenograft growth in vivo. Orally bioavailable.
  • Chemical Name: 5-[[(5-Chloro-2-methyl-3-pyridinyl)amino]methyl]-N-[(1S)-1-(cyclopentylmethyl)-2-(cycloprpylamino)-2-oxoethyl]-2-thiophenecarboxamide
  • Formula: C23H29ClN4O2S
  • CAS Number: 1404456-53-6
  • Solubility: Soluble to 100mM in DMSO and to 50mM in ethanol
  • Purity: >98
Description & Specifications

Specifications

GSK 2830371
>98
50mg
GSK 2830371
461.02
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit