Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GSK-3 alpha/beta Rabbit anti-Human, Mouse, Clone: L5, Novus Biologicals™
Rabbit Monoclonal Antibody
Specifications
Antigen | GSK-3 alpha/beta |
---|---|
Clone | L5 |
Dilution | Western Blot 1:100-1:2000, Immunoprecipitation 1:10-1:500 |
Applications | Western Blot, Immunoprecipitation |
Classification | Monoclonal |
Description
GSK-3 alpha/beta Monoclonal specifically detects GSK-3 alpha/beta in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
GSK-3 alpha/beta | |
Western Blot 1:100-1:2000, Immunoprecipitation 1:10-1:500 | |
Monoclonal | |
Rabbit | |
Cell Biology, Hypoxia, Phospho Specific, Protein Kinase, Signal Transduction, Stem Cell Markers | |
No buffer with No Preservative | |
DKFZp686D0638, EC 2.7.11, EC 2.7.11.26, glycogen synthase kinase 3 alpha, glycogen synthase kinase-3 alpha, GSK-3 alpha | |
GSK3A | |
IgG | |
Affinity Purified | |
GSK3 alpha and beta 46kDa(beta)and51kDa(alpha) weight may be detected. |
L5 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
RUO | |
Human, Mouse | |
P49840 | |
2931 | |
N-terminal 6His-tagged recombinant full length human GSK3Beta. | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title