Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSK-3 beta (p Ser9) Rat anti-Human, Biotin, Clone: MM0748-19G29, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236787B

Catalog No. NBP236787B


Add to cart

Description

Description

GSK-3 beta p Ser9 Monoclonal antibody specifically detects GSK-3 beta p Ser9 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

GSK-3 beta (p Ser9)
MM0748-19G29
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
GSK3B
This antibody was produced from a hybridoma (rat myeloma fused with spleen cells from a rat immunized with Phosphopeptides containing human GSK-3beta S9 site).
Protein G purified
Cancer, Hypoxia, Lipid and Metabolism, mTOR Pathway, Phospho Specific, Protein Kinase, Signal Transduction, Wnt Signaling Pathway
Primary
2932
Human
Western Blot, Immunocytochemistry, Immunofluorescence
Biotin
PBS with 0.05% Sodium Azide
EC 2.7.11, EC 2.7.11.26, glycogen synthase kinase 3 beta, glycogen synthase kinase-3 beta, GSK-3 beta, GSK3beta isoform
Rat
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
This antibody recognizes human phosphor-S9 GSK 3beta in direct ELISAs and WBs.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit