Learn More
GSK-3 beta (p Ser9) Rat anti-Human, DyLight 350, Clone: MM0748-19G29, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236787UV
Catalog No. NBP236787UV
Add to cart
Description
Description
GSK-3 beta p Ser9 Monoclonal antibody specifically detects GSK-3 beta p Ser9 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
GSK-3 beta (p Ser9)
MM0748-19G29
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Purified
GSK3B
This antibody was produced from a hybridoma (rat myeloma fused with spleen cells from a rat immunized with Phosphopeptides containing human GSK-3beta S9 site).
Protein G purified
Cancer, Hypoxia, Lipid and Metabolism, mTOR Pathway, Phospho Specific, Protein Kinase, Signal Transduction, Wnt Signaling Pathway
Primary
2932
Human
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
EC 2.7.11, EC 2.7.11.26, glycogen synthase kinase 3 beta, glycogen synthase kinase-3 beta, GSK-3 beta, GSK3beta isoform
Rat
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
This antibody recognizes human phosphor-S9 GSK 3beta in direct ELISAs and WBs.