Learn More
GSK-3 beta Rat anti-Human, Alexa Fluor™ 532, Clone: MM0748-19G29, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP236787AF532
Catalog No. NP236787A32
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
GSK-3 beta Monoclonal antibody specifically detects GSK-3 beta in Human samples. It is validated for Western Blot,Flow Cytometry,Immunocytochemistry,Immunofluorescence.
Specifications
Specifications
GSK-3 beta
MM0748-19G29
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
EC 2.7.11, EC 2.7.11.26, glycogen synthase kinase 3 beta, glycogen synthase kinase-3 beta, GSK-3 beta, GSK3beta isoform
Rat
IgG1
Store at 4°C in the dark.
Monoclonal
This antibody recognizes human phosphor-S9 GSK 3beta in direct ELISAs and WBs.
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
GSK3B
This antibody was produced from a hybridoma (rat myeloma fused with spleen cells from a rat immunized with Phosphopeptides containing human GSK-3beta S9 site).
0.1 mL
Primary
2932
Human