Learn More
Enzo Life SciencesSupplier Diversity Partner GSK 3 beta (pSer9) monoclonal antibody (3A8) (100 µg)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADI905761100
Clone: 3A8. Host: Mouse. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to a portion of GSK-3β phosphorylated at Ser9. UniProt: P49841. Species Reactivity: Human, Mouse, Rat, Dog. Applications: ELISA, WB. Purity: Thiophilic adsorption and size exclusion chromatography purified. Formulation: Lyophilized from 1ml of 2x PBS containing 0.09% sodium azide, PEG, and sucrose. Handling: Avoid freeze/thaw cycles. After reconstitution, prepare aliquots and store at -80°C.
Catalog No. 50-658-521