Learn More
Alfa Aesar™ GSK-3beta Peptide Inhibitor
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J66645EXD
 View more versions of this product
Catalog No. AAJ66645EXD
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
1197.23
0.5mg
Lys-Glu-Ala-Pro-Pro-Ala-Pro-Pro-Gln-Ser(PO3H)-Pro-NH2
-30° to -10°C
HPLC
Lyophilized powder
RUO
GSK-3beta Peptide Inhibitor