Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ GSK 4112
Selective Rev-Erbα agonist
$243.60 - $1021.96
Specifications
Purity | >98% |
---|---|
Gene Alias | SR 6452 |
Product Type | GSK 4112 |
Molecular Weight (g/mol) | 396.89 |
Inhibitors | GSK 4112 |
Description
Selective Rev-Erbα agonist (EC50 = 250 nM). Enhances recruitment of nuclear receptor co-repressor (NCoR) peptide to Rev-Erbα; causes acute suppression of Bmal1 gene transcription. Induces phase shifts in the circadian clock.Specifications
>98% | |
GSK 4112 | |
GSK 4112 |
SR 6452 | |
396.89 |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title