Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ GSK 5959
Potent and selective BRPF1 inhibitor
Specifications
Purity | >99% |
---|---|
Product Type | GSK 5959 |
Molecular Weight (g/mol) | 394.47 |
Inhibitors | GSK 5959 |
Description
Potent and selective BRPF1 bromodomain inhibitor (IC50 = 80 nM). Exhibits >100-fold selectivity for BRPF1 over a panel of 35 other bromodomains, including BRPF2/3 and BET family bromodomains. Inhibits BRPF1 interaction with histone H3.3. Cell permeable.Specifications
>99% | |
GSK 5959 | |
394.47 | |
GSK 5959 |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title