Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GSK 789472 hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Selective dopamine D3 receptor antagonist and D2 partial agonist (pEC50 values are <5.5 and 8.3 respectively). Selective against the D4 receptor in both agonist and antagonist assays. Brain penetrant.
Specifications
Quantity | 10 mg |
Formula Weight | 295.81 |
Percent Purity | >99% |
Chemical Name or Material | GSK 789472 hydrochloride |
Safety and Handling
Recommended Storage | Desiccate at Room Temperature |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction