Learn More
Azzota Corp GSK-923295 DFS Item
Manufacturer:  Azzota Corp C1163
GSK-923295 is a novel antimitotic inhibitor of centromere-associated protein E (CENP-E) with potential anticancer activity. Please request if need more quantities
Catalog No. 50-161-3120