GSK J5, Tocris™

Inactive isomer of GSK-J4; also cell permeable ester derivative of the inactive control, GSK-J2

Manufacturer: R&D Systems 4689/10

Quantity 10mg

 View more versions of this product

Catalog No. 46-891-0

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1394854-51-3
    Physical Form Powder
    Assay Percent Range >98%
    Molecular Formula C24H27N5O2
    Formula Weight 417.5
    Chemical Name or Material GSK J5

    Specifications

    Physical Form Powder
    Formula Weight 417.5
    Color Off-white
    Quantity 10mg
    Assay Percent Range >98%
    Molecular Formula C24H27N5O2
    CAS 1394854-51-3
    Recommended Storage Store at RT
    Chemical Name or Material GSK J5

    Store at RT

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit