GSK2578215A, Tocris™

Potent, selective LRRK2 inhibitor; brain penetrant

Manufacturer: R&D Systems 4629/10

Quantity 10mg

 View more versions of this product

Catalog No. 46-291-0

  Check Availability
Add to Cart

    Chemical Identifiers

    Assay Percent Range >99%
    Molecular Formula C24H18FN3O2
    Formula Weight 399.42
    Synonym 5-(2-Fluoro-4-pyridinyl)-2-(phenylmethoxy)-N-3-pyridinylbenzamide
    Chemical Name or Material GSK2578215A

    Specifications

    Formula Weight 399.42
    Quantity 10mg
    Assay Percent Range >99%
    Molecular Formula C24H18FN3O2
    CAS 1285515-21-0
    Synonym 5-(2-Fluoro-4-pyridinyl)-2-(phenylmethoxy)-N-3-pyridinylbenzamide
    Recommended Storage Store at Room Temperature
    Chemical Name or Material GSK2578215A

    Store at Room Temperature

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit