Learn More
Abnova™ GSK3A (Human) Recombinant Protein
For use with Functional Study.
Supplier: Abnova™ P6500
Description
Human GSK3A (NP_063937.2, 1 a.a. - 483 a.a.) full length recombinant protein with GST-tag at N-terminal using baculovirus expression system.Specifications
Functional Study | |
glycogen synthase kinase 3 alpha | |
5 μg | |
DKFZp686D0638 |
2931 | |
The purity was assessed by SDS-PAGE/CBB staining. | |
Stored at -80°C. Aliquot to avoid repeated freezing and thawing. |