Learn More
GSK3B Rabbit anti-Human, Unconjugated, Clone: IFC-7, Abnova™
Rabbit Monoclonal Antibody.
Manufacturer:  Abnova Corporation MAB20522
Catalog No. 89-336-922
Add to cart
Description
Description
GSK3B Monoclonal antibody specifically detects GSK3B in Human samples. It is validated for ICC, WB-Ce, IHC-P, IF.
Specifications
Specifications
GSK3B
IFC-7
glycogen synthase kinase 3 beta
Liquid
A synthetic phosphopeptide corresponding to residues surrounding Ser9 of human GSK3B
100 μg
Primary
2932
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
GSK3B
Rabbit
IgG
Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing.
Monoclonal
Human