Learn More
Aviva Systems Biology Corp GSN Antibody - C-terminal region (ARP54299_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP54299_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GSN. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human GSN. Alternate names: DKFZp313L0718, ADF, AGELApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-61166