Learn More
GSN Mouse anti-Human, Clone: GEL-42, Abnova™
Mouse monoclonal antibody raised against human GSN.
Manufacturer:  Abnova Corporation MAB12861
Catalog No. 89-934-208
Add to cart
Description
Description
The protein encoded by this gene binds to the “plus” ends of actin monomers and filaments to prevent monomer exchange. The encoded calcium-regulated protein functions in both assembly and disassembly of actin filaments. Defects in this gene are a cause of familial amyloidosis Finnish type (FAF). Multiple transcript variants encoding several different isoforms have been found for this gene. [provided by RefSeq]
Specifications
Specifications
GSN
GEL-42
Mouse monoclonal antibody raised against human GSN.
In 1.2% C
DKFZp313L0718
Mouse
IgG1
50 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunohistochemistry (Frozen sections) (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-10 ug/mL) Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
GSN
Human plasma.
Affinity purification
RUO
Primary
2934