Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSPT2 Mouse anti-Human, PE/Cy5.5, Clone: OTI2G9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271768PECY55

Catalog No. N271768PEC5


Add to cart

Description

Description

GSPT2 Monoclonal antibody specifically detects GSPT2 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

GSPT2
OTI2G9
Flow Cytometry
Affinity Purified, Conjugated
GSPT2
Full length recombinant protein of human GSPT2 (NP_060564) produced in HEK293T cells.
Immunogen affinity purified
Core ESC Like Genes, Stem Cell Markers
Primary
23708.0
Flow Cytometry
PE-Cyanine5.5
PBS with 0.05% Sodium Azide
eRF3b, ERF3BGST2, eukaryotic peptide chain release factor GTP-binding subunit ERF3B, Eukaryotic peptide chain release factor subunit 3b, FLJ10441, G1 to S phase transition 2, G1 to S phase transition protein 2 homolog
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit