Learn More
Abnova™ GSR 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00002936T01
Catalog No. 89-946-705
Add to cart
Specifications
Specifications
293T
NM_000637.2
pCMV-GSR full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
GSR 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 56.3kDa
Transient overexpression cell lysate was tested with anti-GSR antibody (Mfr. No. H00002936-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human