Learn More
GSS Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA549598
Catalog No. PA549598
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Predicted to react with mouse, rat, monkey and bovine based on sequence homology.
Glutathione is important for a variety of biological functions, including protection of cells from oxidative damage by free radicals, detoxification of xenobiotics, and membrane transport. The protein encoded by this gene functions as a homodimer to catalyze the second step of glutathione biosynthesis, which is the ATP-dependent conversion of gamma-L-glutamyl-L-cysteine to glutathione. Defects in this gene are a cause of glutathione synthetase deficiency. [provided by RefSeq, Jul 2008]
Specifications
Specifications
GSS
Polyclonal
Unconjugated
GSS
Liquid
GSS
KLH conjugated synthetic peptide between 372-400 amino acids from the C-terminal region of human GSS.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
P48637
Glutathione synthetase, GSH synthetase, GSH-S, Glutathione synthase, GSS
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
2937