Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSS Mouse anti-Canine, Human, Rat, Clone: OTI2F2, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525292

Catalog No. PIMA525292


Add to cart

Description

Description

The protein encoded by this gene is a membrane glycosylphosphatidylinositol-anchored glycoprotein that tends to aggregate into rod-like structures. The encoded protein contains a highly unstable region of five tandem octapeptide repeats. This gene is found on chromosome 20, approximately 20 kbp upstream of a gene which encodes a biochemically and structurally similar protein to the one encoded by this gene. Mutations in the repeat region as well as elsewhere in this gene have been associated with Creutzfeldt-Jakob disease, fatal familial insomnia, Gerstmann-Straussler disease, Huntington disease-like 1, and kuru. Alternative splicing results in multiple transcript variants encoding the same protein.
Specifications

Specifications

GSS
Monoclonal
0.7 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P46413, P48637
Glutathione synthase, Glutathione synthetase, GSH S, GSH synthetase, GSH-S, GSHB, GSHS, GSS, MGC14098, OTTHUMP00000030711
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
25458, 2937, 442962
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI2F2
Unconjugated
GSS
Liquid
Gss
Full length human recombit protein of GSS produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit