Learn More
BioVision IncSupplier Diversity Partner GST Fluorometric Activity Assay Kit: Detect GST Activity in Crude Cell Lysate or Purified Protein Fraction and Detect or Quantitate GST-tagged fusion recombinant protein. Simple, Fluorescence-based in vitro Assay. Detection Sensitivity < 0.5 mU. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K260100
GST Fluorometric Activity Assay Kit: Glutathione S-transferase (GST) is a family of enzymes that play an important role in detoxification of xenobiotics. GST catalyzes the formation of the thiol group of glutathione to electrophilic xenobiotics. It utilizes glutathione to scavenge potentially toxic compounds including those produced as a result of oxidative stress and is part of the defense mechanism against the mutagenic, carcinogenic and toxic effects of such compounds. The assay utilizes monochlorobimane (MCB), a dye that reacts with glutathione. The free form of MCB is almost nonfluorescent, whereas the dye fluoresces blue (ex/em = 380/461 nm) when reacts with glutathione. GST catalyzes the MCB-glutathione reactions and the fluorescence levels are proportionally to the amounts of GST presence in the reaction. The kit can detect GST activity in crude cell lysate or purified protein fraction, and also quantitate GST-tagged fusion protein. Detects < 0.5 mU.
Catalog No. 50-111-9658