Learn More
GST Epitope Tag Mouse anti-Human, Alexa Fluor 405, Clone: 3G10, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NB600446AF405
Catalog No. NBX446AF405
Add to cart
Description
Description
GST Epitope Tag Monoclonal antibody specifically detects GST Epitope Tag in Human samples. It is validated for Western Blot.
Specifications
Specifications
GST Epitope Tag
3G10
Western Blot
Purified
GSTM1
GST protein of the parasitic helminth Schistosoma japonicum expressed in E. coli
Protein G purified
Asthma, Cancer, Epitope Tags, GST Epitope Tags, Immunology
Primary
2944
Human
Western Blot
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
EC 2.5.1.18, glutathione S-alkyltransferase, glutathione S-aralkyltransferase, glutathione S-aryltransferase, glutathione S-transferase M1, glutathione S-transferase mu 1, GST class-mu 1, GST HB subunit 4, GST1, GSTM1-1, GSTM1a-1a, GSTM1b-1b, GTH4, GTM1, H-B, HB subunit 4, MGC26563, MU, MU-1, S-(hydroxyalkyl)glutathione lyase
Mouse
IgG1
100 ul
Store at 4C in the dark.
Monoclonal
Recognizes Glutathione S-Transferase (GST), a 26-kDa protein encoded by the parasitic helminth Schistosoma japonicum.