Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GST Epitope Tag Mouse anti-Mouse, Non-species specific, Alexa Fluor™ 594, Clone: 8-326, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP237824AF594

Catalog No. NP237824A94


Add to cart

Description

Description

GST Epitope Tag Monoclonal antibody specifically detects GST Epitope Tag in Mouse,Non-species specific samples. It is validated for Western Blot.
Specifications

Specifications

GST Epitope Tag
8-326
Western Blot
EC 2.5.1.18, glutathione S-alkyltransferase, glutathione S-aralkyltransferase, glutathione S-aryltransferase, glutathione S-transferase M1, glutathione S-transferase mu 1, GST class-mu 1, GST HB subunit 4, GST1, GSTM1-1, GSTM1a-1a, GSTM1b-1b, GTH4, GTM1, H-B, HB subunit 4, MGC26563, MU, MU-1, S-(hydroxyalkyl)glutathione lyase
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Mouse, Nonspecies specific
Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
GSTM1
Recombinant purified full-length GST
0.1 mL
Primary
2944
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit