Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GST Mouse anti-Tag, Clone: OTI4G1, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA150102

Description
GST (Glutathione S-transferase) is a 26 kDa protein, present in eukaryotes and in prokaryotes, where they catalyze a variety of reactions. Glutathione Stransferase is used to create the GST gene fusion system. This GST-fusion protein can then be purified from cells via its high affinity for glutathione. Fusion Proteins offer an important biological assay for direct protein-to-protein interactions. The GST gene fusion system has been introduced into numerous expression vectors.Specifications
GST | |
Monoclonal | |
0.13 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Mouse | |
Protein G | |
RUO | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Tag |
Western Blot | |
OTI4G1 | |
Unconjugated | |
ANTI GST, ANTI-GST, GST ANTIBODY, GST AB, ANTIBODY FOR GST | |
Recombit GST produced in E.coli | |
100 μL | |
Primary | |
Antibody | |
IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction