Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GST Rabbit anti-All, Unconjugated, Clone: 5B8, MilliporeSigma™

Rabbit Monoclonal Antibody

Specifications

Antigen GST
Applications Dot Blot, ELISA, Immunocytochemistry, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ZRB122325MI
View Documents
MilliporeSigma
ZRB122325UL
25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
ZRB12234XMI
View Documents
MilliporeSigma
ZRB12234X25UL
4 x 25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

GST Monoclonal antibody specifically detects GST in All samples. It is validated for Dot Blot, ELISA, Immunocytochemistry, Western Blotting.
Specifications

Specifications

GST
Unconjugated
Rabbit
Glutathione S-transferase
Full-length recombinant E. Coli Glutathione S-transferase.
Primary
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Dot Blot, ELISA, Immunocytochemistry, Western Blot
Purified
All
Gst
Monoclonal
Protein A Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit