Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ GST Tag Monoclonal Antibody,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ 710974
Description
The GST Tag™ Monoclonal Antibody is a mouse monoclonal antibody (IgG1) with high affinity for the 26kDa glutathione-S-transferase (GST) domain from Schistosoma japonicum. This highly purified antibody is used for detection of fusion proteins containing the GST Tag expressed with the pET-41 or pET-42 vector series or other GST-encoding vectors. The 50μg package size provides enough purified antibody to perform 50 Western blots (10 cm x 10 cm).glutathione-S-transferase (GST) domain from Schistosoma japonicum. This highly purified antibody is used for detection of fusion proteins containing the GST Tag expressed with the pET-41 or pET-42 vector series or other GST-encoding vectors.Specifications
GST Tag | |
Monoclonal | |
Stabilized solution in 50% glycerol | |
250 μg | |
Primary | |
-20°C | |
IgG1 |
Immunofluorescence, Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
RUO | |
220aa GST Protein, precise epitope not determined | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?
Provide Content Correction