Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ GST Tag Monoclonal Antibody,
Mouse Monoclonal Antibody
$366.63 - $1267.24
Specifications
Antigen | GST Tag |
---|---|
Applications | Immunofluorescence, Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Conjugate | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Conjugate | Quantity | Price | Quantity & Availability | ||||
71-097-3MI
|
MilliporeSigma™
710973 |
Unconjugated | 50 μg |
Each for $366.63
|
|
||||
71-097-4MI
|
MilliporeSigma™
710974 |
Unconjugated | 250 μg |
Each for $1,267.24
|
|
||||
Description
The GST Tag™ Monoclonal Antibody is a mouse monoclonal antibody (IgG1) with high affinity for the 26kDa glutathione-S-transferase (GST) domain from Schistosoma japonicum. This highly purified antibody is used for detection of fusion proteins containing the GST Tag expressed with the pET-41 or pET-42 vector series or other GST-encoding vectors. The 50μg package size provides enough purified antibody to perform 50 Western blots (10 cm x 10 cm).glutathione-S-transferase (GST) domain from Schistosoma japonicum. This highly purified antibody is used for detection of fusion proteins containing the GST Tag expressed with the pET-41 or pET-42 vector series or other GST-encoding vectors.Specifications
GST Tag | |
Monoclonal | |
Liquid | |
RUO | |
IgG1 | |
-20°C |
Immunofluorescence, Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Stabilized solution in 50% glycerol | |
Primary | |
220aa GST Protein, precise epitope not determined |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title