Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GST Tag Mouse anti-Tag, Clone: 3G12B10, Proteintech
SDP

Catalog No. 501736444 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6444 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6444 Supplier Proteintech Group Inc Supplier No. 660012IG150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

This antibody recognizes the GST-tag fused to either the N- or C-terminus of targeted proteins.

GST (Glutathione S-transferase) is a 26 kDa protein, present in eukaryotes and in prokaryotes, where they catalyze a variety of reactions. Glutathione Stransferase is used to create the GST gene fusion system. This GST-fusion protein can then be purified from cells via its high affinity for glutathione. Fusion Proteins offer an important biological assay for direct protein-to-protein interactions. The GST gene fusion system has been introduced into numerous expression vectors.

Specifications

Antigen GST Tag
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 3G12B10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene Alias GST, GST Tag
Host Species Mouse
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.