Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GSTA3 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP153196
Description
GSTA3 Polyclonal specifically detects GSTA3 in Human samples. It is validated for Western Blot.Specifications
GSTA3 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
Q16772 | |
GSTA3 | |
Synthetic peptides corresponding to GSTA3(glutathione S-transferase alpha 3) The peptide sequence was selected from the middle region of GSTA3. Peptide sequence SSLISNFPLLKALKTRISNLPTVKKFLQPGSPRKPPADAKALEEARKIFR. | |
Affinity Purified | |
RUO | |
2940 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
EC 2.5.1.18, glutathione S-alkyltransferase A3, glutathione S-aralkyltransferase A3, glutathione S-aryltransferase A3, glutathione S-transferase A3, Glutathione S-transferase A3-3, glutathione S-transferase alpha 3, GST class-alpha member 3, GSTA3-3, GTA3, MGC22232, S-(hydroxyalkyl)glutathione lyase A3 | |
Rabbit | |
25 kDa | |
100 μL | |
Primary | |
Mouse: 79%; Sheep: 77%. | |
Human, Mouse, Sheep | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title