Learn More
Abnova™ GSTA4 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00002941T02
Catalog No. 89-946-722
Add to cart
Specifications
Specifications
293T
NM_001512.2
pCMV-GSTA4 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
GSTA4 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 24.53kDa
Transient overexpression cell lysate was tested with anti-GSTA4 antibody (Mfr. No. H00002941-B02) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human