Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSTA4 Mouse anti-Human, Alexa Fluor™ 647, Clone: OTI3F5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270856AF647

Catalog No. N270856A647


Add to cart

Description

Description

GSTA4 Monoclonal antibody specifically detects GSTA4 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications

Specifications

GSTA4
OTI3F5
Western Blot, Flow Cytometry, Immunohistochemistry
Affinity Purified, Conjugated
GSTA4
Full length recombinant protein of human GSTA4 (NP_001503) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Flow Cytometry, Immunohistochemistry, Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
DKFZp686D21185, EC 2.5.1.18, glutathione S-alkyltransferase A4, glutathione S-aralkyltransferase A4, glutathione S-aryltransferase A4, glutathione S-transferase A4, Glutathione S-transferase A4-4, glutathione S-transferase alpha 4, glutathione transferase A4-4, GST class-alpha member 4, GSTA4-4, GTA4, S-(hydroxyalkyl)glutathione lyase A4
Mouse
IgG1
0.1 mL
Primary
2941.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit