Learn More
Abnova™ GSTA5 293T Cell Transient Overexpression Lysate (Denatured) (T02)
Human overexpression lysate
Manufacturer:  Abnova™ H00221357T02
Catalog No. 89-946-726
Add to cart
Specifications
Specifications
293T
BC148816
pCMV-GSTA5 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
GSTA5 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 24.42kDa
Transient overexpression cell lysate was tested with anti-GSTA5 antibody (Mfr. No. H00221357-D01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human