Learn More
GSTM2 Rabbit anti-Human, DyLight 350, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP299918UV
Catalog No. NB063505
Add to cart
Description
Description
GSTM2 Polyclonal antibody specifically detects GSTM2 in Human samples. It is validated for Western Blot, ELISA
Specifications
Specifications
GSTM2
DyLight 350
50mM Sodium Borate
glutathione S-transferase mu 2 (muscle), MGC117303, muscle, S-(hydroxyalkyl)glutathione lyase M2
Produced in rabbits immunized with purified, recombinant GSTM2 (P28161; Met 1-Lys 218).
Affinity purified
RUO
Primary
2946
ELISA, Western Blot
Western Blot, ELISA
Purified
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Polyclonal
Human