Learn More
Aviva Systems Biology Corp GSTM3 Antibody - middle region (ARP53562_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP53562_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GSTM3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human GSTM3. Alternate names: GST5, GSTB, GSTM3-3, GTM3, MGC3310, MGC3704ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-61050