Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSTM4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549332

Catalog No. PA549332

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Conjugation of reduced glutathione to a wide number of exogenous and endogenous hydrophobic electrophiles. Active on 1- chloro-2,4-dinitrobenzene.
Specifications

Specifications

GSTM4
Polyclonal
PBS with 0.09% sodium azide
Q03013
Glutathione S-transferase Mu 4, GST class-mu 4, GST-Mu2, GSTM4-4, GSTM4
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
2948
Western Blot
Unconjugated
GSTM4
Liquid
GSTM4
KLH conjugated synthetic peptide between 16-50 amino acids from the N-terminal region of human GSTM4.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit