Learn More
GSTP1 Rabbit anti-Human, Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Manufacturer:  Invitrogen MA527913
Catalog No. PIMA527913
Add to cart
Description
Description
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using Recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Glutathione S-transferases (GSTs) are a family of enzymes that play an important role in detoxification by catalyzing the conjugation of many hydrophobic and electrophilic compounds with reduced glutathione. Based on their biochemical, immunologic, and structural properties, the soluble GSTs are categorized into 4 main classes: alpha, mu, pi, and theta. This GST family member is a polymorphic gene encoding active, functionally different GSTP1 variant proteins that are thought to function in xenobiotic metabolism and play a role in susceptibility to cancer, and other diseases.
Specifications
Specifications
GSTP1
Recombinant Monoclonal
Unconjugated
GSTP1
Liquid
Rabbit
IgG
100 μl
-20°C
Primary
2950
Immunohistochemistry, Western Blot
RM347
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide
P09211
GSTP1
A peptide corresponding to the C-terminus of human GST3/GST-pi
Protein A
RUO
Antibody
Monoclonal
Human