Learn More
GSTP1 Rabbit anti-Human, Unconjugated, Clone: DD-7, Abnova™
Rabbit Monoclonal Antibody.
Manufacturer:  Abnova Corporation MAB20113
Catalog No. 89-336-513
Add to cart
Description
Description
GSTP1 Monoclonal antibody specifically detects GSTP1 in Human samples. It is validated for WB, IP, ICC.
Specifications
Specifications
GSTP1
DD-7
glutathione S-transferase pi 1
Liquid
Rabbit
IgG
Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing.
Monoclonal
Human
Immunocytochemistry, Immunoprecipitation, Western Blot
Unconjugated
GSTP1
DFN7, FAEES3, GST3, PI
A synthetic peptide corresponding to GST
100 μg
Primary
2950