Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare GSTrap™ FF Columns: 7 × 25mm: PROMO

Family

Manufacturer:  GE Healthcare 17513001PM

Catalog No. 45-000-281LR

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

  • Mild elution conditions preserve antigenicity and function of proteins extracted
  • Prepacked with Glutathione Sepharose™ media
  • Average particle size: 90μm
  • Bed dimensions: 7 × 25mm
  • Flow velocity: <4mL/min.
Media
  • Matrix: Highly cross-linked 4% agarose
  • Ligand: glutathione and 10-carbon linker arm
  • Binding capacity: >10mg/mL
  • pH stability working range: 3 to 12

Specifications

Specifications

1mL
7mm
Glutathione and 10 carbon linker arm
5 × 1mL
90μm
3 bar
4° to 30°C
0.27 in.
<4mL/min.
Polypropylene
3 to 12
GSTrap column
5 × 1mL
4% Highly Cross-linked Agarose
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit