Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare GSTrap™ HP Columns PROMO

Use to provide high-resolution, one-step purifications of glutathione S-Transferase (GST) tagged proteins. GE Healthcare Life Sciences™ GSTrap™ HP Columns are prepacked with Glutathione Sepharose™ High Performance medium.

Manufacturer:  GE Healthcare 29361662

Catalog No. 45-000-333PM

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

  • One-step purification of GST-tagged proteins and other glutathione S-transferases or glutathione-dependent proteins
  • Glutathione coupled to Sepharose™ High Performance media
  • Column material: polypropylene
  • May be used with syringe, peristaltic pump or chromatography system
  • Purification process preserves antigenicity and function
  • Available as 1mL and 5mL columns
Media
  • Average particle size: 34µm
  • Ligand: Glutathione
  • Matrix: Highly cross-linked 6% agarose
  • Binding capacity: >7mg/mL
  • pH stability working range: 3 to 12

1mL columns

  • Bed dimensions: 7 x 25mm
  • Flow velocity: <4mL/min.
5mL columns
  • Bed dimensions: 16 x 25mm
  • Flow velocity: <20mL/min.

Specifications

Specifications

5mL
16mm
Glutathione
1 x 5mL
34μm
3 bar
4° to 30°C
0.62 in.
<20mL/min.
Polypropylene
3 to 12
GSTrap HP column
1 x 5mL
4% Highly Cross-linked Agarose
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit