Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GSTT1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543186
Description
Peptide sequence: TWRQRVEAAV GEDLFQEAHE VILKAKDFPP ADPTIKQKLM PWVLAMIR Sequence homology: Cow: 79%; Dog: 86%; Guinea Pig: 79%; Horse: 86%; Human: 100%; Rabbit: 79%; Rat: 86%.
Glutathione S-transferase (GST) theta 1 (GSTT1) is a member of a superfamily of proteins that catalyze the conjugation of reduced glutathione to a variety of electrophilic and hydrophobic compounds. Human GSTs can be divided into five main classes: alpha, mu, pi, theta, and zeta. The theta class includes GSTT1 and GSTT2. The GSTT1 and GSTT2 share 55% amino acid sequence identity and both of them were claimed to have an important role in human carcinogenesis. The GSTT1 gene is located approximately 50kb away from the GSTT2 gene. The GSTT1 and GSTT2 genes have a similar structure, being composed of five exons with identical exon/intron boundaries.
Specifications
GSTT1 | |
Polyclonal | |
Unconjugated | |
Gstt1 | |
AI255817; Glutathione S-transferase 1 (theta); glutathione S-transferase 5; glutathione S-transferase theta 1; glutathione S-transferase theta-1; glutathione S-transferase, theta 1; glutathione transferase T1-1; GST 5-5; GST class-theta-1; GSTT1; Gstt1-1; GSTYRS; similar to Glutathione S-transferase theta 1 (GST class-theta 1) (Glutathione transferase T1-1) | |
Rabbit | |
Affinity Chromatography | |
RUO | |
2952 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P30711 | |
Gstt1 | |
Synthetic peptide directed towards the C-terminal of human GSTT1 (aa 193-240). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction